Learn More
Thermo Scientific™ KingFisher Duo Prime Accessories
Product Code. 30131520 Shop All Thermo Scientific Products
Change view
Click to view available options
Format:
24 Deep-well Head
24 Deep-well Heating Block
96 Deep-well Head
96 Deep-well Heating Block
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Format Unit Size
30131520 24 Deep-well Heating Block Each
30132313 24 Deep-well Head Each
30131189 96 Deep-well Head Each
30131332 96 Deep-well Heating Block Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 30131520 Supplier Thermo Scientific™ Supplier No. N12462
 Restricted Item
Expédition estimée: 11-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
The KingFisher Duo Prime system is a flexible system for sample processing that offers highly versatile, automated magnetic-particle processing for DNA/RNA, protein, and cell purification from a wide range of sources.
The KingFisher Duo Prime system is a flexible system for sample processing that offers highly versatile, automated magnetic-particle processing for DNA/RNA, protein, and cell purification from a wide range of sources. Using revolutionary magnetic particle separation technology, this system provides excellent reproducibility and quality.
The KingFisher Duo Prime instrument comes with a 12-pin magnet head for 96 deep well applications, a 6-pin magnet head for 24 deep well applications, and heating blocks. Additional magnet heads and heating blocks are offered here for separate purchase.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Nucleic Acid Extraction and Cell Separation Instrument
Sample Type Fresh or Frozen Plant Tissues, FFPE Tissue, Cells, Blood, Plasma
Format 24 Deep-well Heating Block
Capacity 6 Samples
Disposable Reusable
For Use With (Equipment) KingFisher Duo Prime Purification System
Quantity 1 Block
For Research Use Only. Not for use in diagnostic procedures.