Learn More
Thermo Scientific™ KingFisher™ PlasmidPro Maxi Processor Endotoxin-Free Cartridge
Shop All Thermo Scientific ProductsDescripción
The KingFisher™ PlasmidPro Maxi Processor Endotoxin-Free Cartridge is used with the KingFisher™ PlasmidPro Maxi Processor for fully automated isolation of endotoxin-free plasmid DNA in under 75 minutes. Purify high-quality plasmid DNA that is suitable for advanced transfection applications such as transfection of sensitive cells lines or genome engineering.
Features and benefits
- Convenient one-piece cartridge pre-filled with all necessary reagents
- Collection tube included for simple removal and storage of plasmid DNA
- Designed for mess-free clean-up and waste disposal
- Plasmid DNA contains less than <0.1 EU/μg
Additional product details
The KingFisher™ PlasmidPro Maxi Processor is a cutting-edge solution designed for fully automated maxi-scale plasmid purification.
At the end of the procedure, the system delivers endotoxin-free plasmid DNA. The purified DNA is resuspended in TE buffer, ensuring optimal quality and compatibility with downstream applications.
Especificaciones
Especificaciones
| Product Type | Cartridge |
| Content And Storage | 4 KingFisher™ PlasmidPro Maxi Processor Endotoxin-free Cartridges Store at room temperature (15°C to 25°C). |
| For Use With (Application) | Plasmid purification |
| For Use With (Equipment) | KingFisher™ PlasmidPro Maxi Processor |
| Purification Target | Plasmid DNA |
| Quantity | 4 Preps |
| Run Time | <75 min. |
| Sample Volume | 150 mL |
| Shipping Condition | Room Temperature |
Frequently Asked Questions (FAQs)
When using frozen pellets as a starting point, please consider the following guidelines:
- The pellet should represent 150 mL of the specified culture.
- Fully resuspend the pellet in 150 mL of 1X PBS or the parent media.
- The resuspension can be achieved by either pipetting up and down or vigorous vortexing.
We recommend storing the KingFisher PlasmidPro Maxi Processor Cartridge at room temperature. Refrigeration is not needed.
The KingFisher PlasmidPro Maxi Processor Cartridge is single-use and should be disposed of after use, minimizing clean-up and risk of cross-contamination. Re-use of the cartridge will contaminate and potentially harm the instrument.
No. The RNaseA used in the KingFisher PlasmidPro Maxi Processor Cartridge is not animal origin-free. Review the Certificate of Analysis (COA) for more details.
At the end of the purification using the KingFisher PlasmidPro Maxi Processor, the plasmid DNA sample is resuspended in TE buffer (10 mM Tris-HCl, pH 8.0, 0.1 mM EDTA).
For Research Use Only. Not for use in diagnostic procedures.
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.