Learn More
Usbeck Laborgeraete™ Knife
Product Code. 10528301
Change view
Click to view available options
Length (Metric):
185 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
10528301 185 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10528301 Supplier Usbeck Laborgeraete™ Supplier No. 5210
  • ONLINE EXCLUSIVE PRICE
    45.45€
    40.20€ / 1 pièces
    Économisez 5.25€
    11% de réduction
Expédition estimée: 02-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Order Info
LengthMetric
Specifications
Product Type Knife
Handle Type Plastic
Length (Metric) Blade 60 mm
Length (Metric) 185 mm