Learn More
DWK Life Sciences™ Kimble™ Kontes™ Bubbler Tube, Monier-Williams, 24/40 Joint
Shop All DWK Life Sciences ProductsDescription
- Bubbler Tube
- Standard Taper joint 24/40
- Case of 1
- Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Specifications
Specifications
| Type | Bubbler Tube |
| Standard Taper Joint | 24/40 |