Learn More
VELP Scientifica™ KS 1000 Scrubber
BIOPHARMA
Product Code. 17804687 Shop All Velp Products
Change view
Click to view available options
Capacity (Metric):
3.5 L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
17804687 3.5 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Product Code. 17804687 Supplier VELP Scientifica™ Supplier No. F307A0660
  • ONLINE EXCLUSIVE PRICE
    3105.00€
    2694.00€ / 1 pièces
    Économisez 411.00€
    13% de réduction
View all promotions
En stock
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Designed to neutralize toxic fumes produced during mineralization and safeguard laboratory operators without requiring any connection to tap water.
Innovative and compact device that is designed to neutralize toxic fumes produced during mineralization and safeguard lab operators, without requiring any connection to tap water.
  • Perfectly complements VELP's compact semi-automatic and automatic digesters, fitting seamlessly into the fume hood for an optimized working environment.
  • An intuitive user interface, conveniently placed on top of the instrument, simplifies operations.
  • With 4 adjustable suction power levels, laboratory operators have control over their specific requirements.
  • Carefully selected materials ensure durability even in the most demanding conditions with reduced maintenance costs.
  • Tanks made of glass and connection tubes constructed with Viton offer superior performance and increased resistance.
  • The eight punctured discs contribute significantly to achieving maximum neutralization efficiency.
  • The innovative design allows for easy access to all components, facilitating quick and effortless cleaning and maintenance.
  • In specific applications, an optional activated carbon filter can be used to absorb odours produced during oxidative mineralization.
Specifications
Product Type Fume Removal System
Display No Display
Capacity (Metric) 3.5 L
Flow Rate Up to 25 L/min.
For Use With (Equipment) KS 1000 Scrubber
Material Metal structure and PVC components