Learn More
L-Ribose, TRC
Product Code. 30179505
Change view
Click to view available options
Quantity:
1 g
5 g
100 g
Unit Size:
100g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30179505 5 g 5g
30178962 1 g 1g
30179293 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30179505 Supplier Toronto Research Chemicals Supplier No. TRCR4175055G
 Restricted Item
Expédition estimée: 10-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 24259-59-4
Molecular Formula C5 H10 O5
Molecular Weight (g/mol) 150.13
Synonym L-Ribose, Ribose, L- (8CI), L-(+)-Ribose
IUPAC Name (2S,3S,4S)-2,3,4,5-tetrahydroxypentanal
SMILES OC[C@H](O)[C@H](O)[C@H](O)C=O
Specifications
Quantity 5 g
InChI Formula InChI=1S/C5H10O5/c6-1-3(8)5(10)4(9)2-7/h1,3-5,7-10H,2H2/t3-,4+,5-/m1/s1
Formula Weight 150.0528
Chemical Name or Material L-Ribose