Learn More
Miele Professional™ Lab Washer PLW 8636 [LAB] with Liquid Dispensing, DryPlus, Hot Air Drying, and Oil-Resistant Door Seal
Product Code. 30533224 Shop All Miele Products
Change view
Click to view available options
Electrical Requirements:
400 V, 3N, 50 Hz
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Electrical Requirements
30533224 400 V, 3N, 50 Hz
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Product Code. 30533224 Supplier Miele Professional™ Supplier No. 12637620
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
With liquid dispensing and Efficient hot air drying - DryPlus
  • Performance/cycle, e.g. 128 ST laboratory bottles or 98 ST pipettes
  • Comprehensive, fully flexible programming for your requirements
  • Powerful, variable-speed and highly robust pump
  • Drying unit DryPlus for challenging laboratory glassware
  • Flexible and simple - modular load carriers & EasyLoad system
  • Functional enhancement made possible with retrofitting kits
  • Safety through monitoring - wash pressure and spray arm monitoring
  • Efficient hot air drying - DryPlus Connectivity as standard via WiFi or Ethernet
TRUSTED_SUSTAINABILITY
Specifications
Display Full-color Touch Screen
Material Stainless Steel
Product Type Laboratory Washer/Dryer
Flow Rate 600 L/min.
Max. Pressure 1000 kPa
Min. Purified Water Pressure 200 kPa
For Use With (Application) Cleaning of Laboratory Glassware
For Use With (Equipment) Washer-Disinfectors
Includes DryPlus
Inlet Water Temperature (English) Min. 5°C
Inlet Water Temperature (English) Recommended 5°C to 65°C
Inlet Water Temperature (English) Max. 65°C
Inlet Water Temperature (Metric) Max. 65°C
Inlet Water Temperature (Metric) Min. 5°C
Inlet Water Temperature (Metric) Recommended 5°C to 65°C
Depth (English) 27 9/16 in.
Depth (Metric) 70.00 cm
Height (Metric) 143.00 cm
Width (English) 35 7/16 in.
Width (Metric) 90.00 cm
Electrical Requirements 400 V, 3N, 50 Hz
Voltage 400 V
Frequency 50 Hz