Learn More
SHP Steriltechnik AG Laboklav 135 Trolley
Product Code. 31908800
Change view
Click to view available options
Capacity (Metric):
280 kg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
31908800 280 kg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31908800 Supplier SHP Steriltechnik AG Supplier No. 20.0135.000.301
Expédition estimée: 21-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Trolley for Laboklav 135, horizontal unit.
  • Stainless Steel
  • With 4 castors for easy transport
Specifications
Product Type Trolley for Steam Sterilizer
Capacity (Metric) 280 kg
For Use With (Equipment) Laboklav 135