Learn More
Thermo Scientific™ LabTower™ AFT Clinical Water Purification System
Product Code. 16610956 Shop All Thermo Scientific Products
Change view
Click to view available options
Description:
LabTower AFT 20
RO Flow Rate:
20 L/hr.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description RO Flow Rate
16610956 LabTower AFT 20 20 L/hr.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Product Code. 16610956 Supplier Thermo Scientific™ Supplier No. 50135466
  • ONLINE EXCLUSIVE PRICE
    17380.00€
    16302.00€ / 1 pièces
    Économisez 1078.00€
    6% de réduction
 Restricted Item
Expédition estimée: 30-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Ideal to feed clinical analyzer systems, these 20 or 40L/hr systems ship complete with pre-treatment and 100L integrated tank.
Ideal to feed clinical analyzer systems, choose either the Thermo Scientific™ LabTower AFT 20 or 40L/hr version and it will ship complete with pre-treatment and 100L integrated tank.
The system utilizes the following water purification technologies:
  • Pre-treatment by means of a 10inch mark (in.) filter housing with a 5Glc mum filter and hardness stabilizer for protection of downstream purification media
  • Reverse osmosis module, for the removal of organic and inorganic contaminants, micro organisms, particles and colloids
  • Ion Exchanger, for the removal of dissolved organic substances and inorganic ions
  • Digital control, for automatic monitoring and controlling of the reverse osmosis system
  • Storage tank with water level control for fully automatic maintenance of a stock of high purity water that is protected from light
TRUSTED_SUSTAINABILITY
Specifications
Bacterial retention 0.99
Conductivity 0.1 to 1.0 μS/cm
Description LabTower AFT 20
Feedwater Connector R 3/4 in.
Flow Rate 20 L/hr.
Resistivity 10 to 1 MΩ·cm
Operating Pressure 2 to 6 bar
Power Consumption 0.25 kW
RO Flow Rate 20 L/hr.
Temperature Range (Metric) 2°C to 35°C
Width (Metric) 450 mm
Height (Metric) 1500 mm
Depth (Metric) 580 mm
Net Weight (Metric) 66 kg
Electrical Requirements 230 V 50 Hz
Voltage 230 V
Frequency 50 Hz