Learn More
Landiolol hydrochloride, MedChemExpress
Product Code. 16483098
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16483098 10 mg 10mg
16415837 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16483098 Supplier Medchem Express Supplier No. HY100607A
 Restricted Item
Expédition estimée: 08-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
MedChemExpress Landiolol hydrochloride (ONO1101 hydrochloride) is a highly beta1 selective ultra-short acting beta-blocker (β1/β2 selectivity = 255:1, a half-life of 4 min), acts as an adrenoceptor antagonist.
Landiolol hydrochloride (ONO1101 hydrochloride) is a highly beta1 selective ultra-short acting beta-blocker (β1/β2 selectivity = 255:1, a half-life of 4 min), acts as an adrenoceptor antagonist.
Specifications
Chemical Name or Material Landiolol hydrochloride
CAS 144481-98-1
Color White
Purity Grade Notes Research
Molecular Formula C25H40ClN3O8
Quantity 10 mg
Synonym ONO1101 hydrochloride
Solubility Information DMSO : 250 mg/mL (457.83 mM; Need ultrasonic) ∣H2O : ≥ 100 mg/mL (183.13 mM)
SMILES O=C(OC[C@H]1OC(C)(C)OC1)CCC2=CC=C(OC[C@@H](O)CNCCNC(N3CCOCC3)=O)C=C2.[H]Cl
Molecular Weight (g/mol) 546.05
Formula Weight 546.05
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation