Learn More
Levamisol 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Product Code. 30321782
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30321782 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30321782 Supplier Dr. Ehrenstorfer Supplier No. DREA14629690AL100
 Restricted Item
Expédition estimée: 07-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Levamisol
CAS 14769-73-4
Analyte or Component Names Levamisol
Shipping Condition Room Temperature
Molecular Formula C11 H12 N2 S
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
Synonym Imidazo[2,1-b]thiazole, 2,3,5,6-tetrahydro-6-phenyl-, (S)-, (6S)-2,3,5,6-Tetrahydro-6-phenylimidazo[2,1-b]thiazole, (-)-2,3,5,6-Tetrahydro-6-phenylimidazo[2.1-b]thiazole, (-)-Tetramisole, (S)-(-)-Levamisole, 5: PN: WO2015086738 SEQID: 8 claimed sequence, Anthelsol, Ketrax, L-Tetramisole, Lepuron, Levamisol, Levamisole, Levipor, Levomol, Levomysol, Levovermax, Nilverm LV, Totalon, Vermisol 150, Wormicid, l-2,3,5,6-Tetrahydro-6-phenylimidazo[2,1-b]thiazole, l-Tetramisole
InChI Formula InChI=1S/C11H12N2S/c1-2-4-9(5-3-1)10-8-13-6-7-14-11(13)12-10/h1-5,10H,6-8H2/t10-/m1/s1
SMILES C1CN2C[C@@H](N=C2S1)c3ccccc3
IUPAC Name (6S)-6-phenyl-2,3,5,6-tetrahydroimidazo[2,1-b][1,3]thiazole
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 204.29
Quantity 1 mL