Learn More
R&D Systems™ Liarozole hydrochloride
Product Code. 18075202
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18075202 10 mg 10mg
18034632 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18075202 Supplier R&D Systems™ Supplier No. 2705/10
 Restricted Item
Expédition estimée: 27-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
A Tocris product, part of R&D Systems. Blocks retinoic acid metabolism
Cytochrome P450 inhibitor. Inhibits several cytochrome P450 enzymes including aromatase (CYP19) and retinoic acid 4-hydroxylase (CYP26). Blocks retinoic acid metabolism (retinoic acid metabolism blocking agent, RAMBA). Displays antitumor activity against androgen-dependent and independent rat prostate carcinomas.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 145858-50-0
Molecular Formula C17H14Cl2N4
Specifications
Quantity 10 mg
Product Line Tocris
Formula Weight 345.23
Purity >99%
Molecular Weight (g/mol) 345.227
Chemical Name or Material Liarozole hydrochloride