Description
- Basic PC software allows sample measurement only
- Data calculations must be performed separately (e.g., in Excel)
- Sirius 2 license for FB12/Sirius PC software (Microsoft Windows) includes:
- Single Assay protocol
- Dual Assay protocol
- Single Kinetics protocol
- Multiple Kinetics protocols
- Cut-off protocol
- Calculation of replicates
- Blank subtraction
- Additional data processing functions
Specifications
Specifications
| Type | Software |
| For Use With (Application) | Luminescence Assays |
| For Use With (Equipment) | Sirius 2 LB 9526 Tube Luminometer |
| System Requirements | Windows 10 or Newer |