Learn More
Checkers Linebacker™ 5 Channel Heavy Duty Cable Protector
Product Code. 17792418
Change view
Click to view available options
Color:
Yellow, Orange
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
17792418 Yellow, Orange Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17792418 Supplier Checkers Supplier No. PLBS0002YO
  • ONLINE EXCLUSIVE PRICE
    351.13€
    285.24€ / 1 pièces
    Économisez 65.89€
    18% de réduction
Expédition estimée: 26-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Linebacker 5-channel cable protector with hinged lid provides a safer crossing for vehicles and pedestrian traffic while protecting valuable electrical cables and hose lines from damage in extreme weather conditions and work environments.
  • Linebacker™ heavy duty 5-channel cable protector with hinged lid provides a safer crossing for vehicles and pedestrian traffic while protecting valuable electrical cables and hose lines from damage in extreme weather conditions and work environments
  • The modular interlocking design is ideal for use in oil and gas applications, mining, military, and other industrial environments with heavy trucks and forklifts
Specifications
Length (English) 36 in.
Length (Metric) 91 cm
Width (Metric) 51 cm
Color Yellow, Orange
Material Polyurethane
Type Cable Protector