Learn More
Lonafarnib, 98%, BLD Pharm™
Product Code. 30801486
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
50 mg
100 mg
250 mg
Unit Size:
100mg
10mg
1mg
250mg
25mg
50mg
5mg
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30801486 100 mg 100mg
30796649 1 mg 1mg
30798853 5 mg 5mg
30797929 50 mg 50mg
30790626 25 mg 25mg
30796088 10 mg 10mg
30797777 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 30801486 Supplier BLD Pharm Supplier No. BD136650100mg
 Restricted Item
Expédition estimée: 25-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
CAS 193275-84-2
Molecular Formula C27H31Br2ClN4O2
MDL Number MFCD06795138
Quantity 100 mg
SMILES O=C(N1CCC(CC(N2CCC([C@@H]3C4=C(Br)C=C(Cl)C=C4CCC5=CC(Br)=CN=C53)CC2)=O)CC1)N
Molecular Weight (g/mol) 638.82
Percent Purity 98%
Chemical Name or Material Lonafarnib