Learn More
Lonza™ IsoGel™ IEF Plates
Product Code. 11507995
Change view
Click to view available options
pH Range:
3 to 10
6 to 10.5
7 to 11
Unit Size:
Each
Pack of 6
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. pH Range Unit Size
11507995 3 to 10 Pack of 6
11517995 6 to 10.5 Pack of 6
12890125 7 to 11 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11507995 Supplier Lonza™ Supplier No. 56015
  • ONLINE EXCLUSIVE PRICE
    942.00€
    726.67€ / 6 pièces
    Économisez 215.33€
    22% de réduction
 Restricted Item
Expédition estimée: 25-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Precast agarose IEF plates provide rapid and reproducible isoelectric focusing of proteins up to 2000kDa
  • Specifically designed for antibody analysis of
  • Each gel is supported on GelBond™ film for easy handling and dimensional stability
  • Gel size is compatible with most IEF chambers
  • 125 x 100mm
Isoelectric focusing, antibody separation and analysis
Order Info
Accessories packs (Cat. No. BMA56014 and BMA56010) and blotting paper (BMA56007) available separately.
TRUSTED_SUSTAINABILITY
Specifications
pH Range 3 to 10
Separation Type Isoelectric Focusing
Form Solid
For Use With (Application) Isoelectric focusing, Antibody separation and analysis
Storage Requirements 2° to 8°C
Product Type Gel Plate