Learn More
Lovibond™ RD125 Reactor
Product Code. 12681735
Change view
Click to view available options
Material:
Round Metal block in Epoxy covered housing
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Unit Size
12681735 Round Metal block in Epoxy covered housing Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Product Code. 12681735 Supplier Lovibond™ Supplier No. 2418940
  • ONLINE EXCLUSIVE PRICE
    1235.00€
    1109.10€ / 1 pièces
    Économisez 125.90€
    10% de réduction
View all promotions
Expédition estimée: 08-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Used for the chemical digestion of water samples, such as in the determination of COD, TOC, Total Phosphate and Total Nitrogen
  • Required temperatures and reaction times are preprogrammed, and easy to select using the keypad
  • Users can pre-select reaction times of 30, 60 and 120 minutes or can set it to operate continuously
  • Power: 100-240 VAC, 50-60Hz
Specifications
Certifications/Compliance CE, UL, TUV
Dimensions (L x W x H) 10.83 x 6.1 x 3.74 in. (27.51 x 15.49 x 9.49 cm)
Voltage 115/230 V
Height (English) 3.74 in.
Height (Metric) 9.49 cm
Includes RD125 Reactor, 16mm COD vial adapter, instructions
Length (English) 10.83 in.
Length (Metric) 27.51 cm
Material Round Metal block in Epoxy covered housing
Range 0 to 15000 mg/L
Weight (English) 7.9 lb.
Weight (Metric) 3.58 kg
Width (English) 6.1 in.
Width (Metric) 15.49 cm