Learn More
Abcam Luminescent ATP Detection Assay Kit
Product Code. 32120073 Shop All Abcam Products
Change view
Click to view available options
Quantity:
300 Tests
Unit Size:
300 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32120073 300 Tests 300 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32120073 Supplier Abcam Supplier No. ab113849300Test
 Restricted Item
Expédition estimée: 18-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Detect Adenosine-5'-triphosphate in 30m in Cell Samples with Luminescent ATP Detection Assay Kit AB113849. Cited in 284 publications. Use with Microplate reader.
Detect Adenosine-5'-triphosphate in 30m in Cell Samples with Luminescent ATP Detection Assay Kit AB113849. Cited in 284 publications. Use with Microplate reader.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Quantity 300 Tests
Regulatory Status RUO