Learn More
Brady Magicard™ CR80 PVC Card with Mag Stripe and HoloPatch
Product Code. 30067369 Shop All Brady Products
Change view
Click to view available options
Color:
White
Unit Size:
Pack of 500
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
30067369 White Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30067369 Supplier Brady Supplier No. 322003
  • ONLINE EXCLUSIVE PRICE
    140.00€
    120.00€ / 500 pièces
    Économisez 20.00€
    14% de réduction
En stock
Add to basket
Add to basket
This item is not returnable. View return policy
Plain white PVC Card (CR80) with magnetic stripe and HoloPatch™ for extra visual security.
  • Pre-approved for best quality printing on Magicard Printers.
  • Magnetic stripe can be programmed with Magicard Printers.
  • HoloPatch™ for extra visual security.
Specifications
Quantity 500 Pk.
Product Type Badge
Alternate Part Number M9006-797
Color White
Material PVC
Message Blank
For Use With (Application) Healthcare and Hospital labelling;General identification;Facility identification;Security identification
Height (English) 3.4 in.
Height (Metric) 86 mm
Thickness 0.762 mm
Width (English) 2.1 in.
Width (Metric) 54 mm
Label Type Cards for Printers
For Use With (Equipment) Magicard 300, Magicard 600, Pronto 100, Prima 8