Learn More
Brady Magicard™ CR80 Rewritable Cards
Product Code. 30067361 Shop All Brady Products
Change view
Click to view available options
Color:
White
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
30067361 White Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30067361 Supplier Brady Supplier No. 322005
  • ONLINE EXCLUSIVE PRICE
    119.00€
    102.00€ / 100 pièces
    Économisez 17.00€
    14% de réduction
En stock
Add to basket
Add to basket
This item is not returnable. View return policy
Plain white PVC Card (CR80).
  • Pre-approved for best quality printing on Magicard Printers.
  • Re-write your card multiple times.
  • Pack of 100 cards.
Specifications
Quantity 100 Pk.
Product Type Badge
Alternate Part Number E9323
Color White
Material PVC
Message Blank
For Use With (Application) Healthcare and Hospital labelling;General identification;Facility identification;Security identification
Height (English) 3.4 in.
Height (Metric) 86 mm
Thickness 0.762 mm
Width (English) 2.1 in.
Width (Metric) 54 mm
Label Type Cards for Printers
For Use With (Equipment) Magicard 300, Magicard 600, Pronto 100, Prima 8