missing translation for 'onlineSavingsMsg'
Learn More

Magnesium bromide hexahydrate, 98+%

Product Code. 11408630
Change view
Click to view available options
Quantity:
100 g
500 g
2 kg
Unit Size:
100g
2kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11408630 2 kg 2kg
11418630 100 g 100g
11428630 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11408630 Supplier Thermo Scientific Alfa Aesar Supplier No. 046120.A3
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13446-53-2 | Br2H12MgO6 | 292.20 g/mol

Magnesium bromide hexahydrate is used as flame retardant. Used as source of magnesium ion and a co-factor for many enzymes including deoxyribonuclease (DNase) and various restriction enzymes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13446-53-2
Molecular Formula Br2H12MgO6
Molecular Weight (g/mol) 292.20
MDL Number MFCD00149780
InChI Key LGLXXNHIGIJYQQ-UHFFFAOYSA-L
Synonym acmc-1bp1b, magnesium bromide-water 1/2/6, magnesium 2+ ion hexahydrate dibromide, magnesium 2+ hexahydrate dibromide, grignard reagent
PubChem CID 17749097
SMILES O.O.O.O.O.O.Br[Mg]Br

Specifications

Melting Point 172°C
Color Colorless
Physical Form Crystalline
Quantity 2 kg
Linear Formula MgBr2·6H2O
Solubility Information Soluble in water, somewhat soluble in alcohol
IUPAC Name dibromomagnesium hexahydrate
Formula Weight 292.22 (184.13 Anhydrous)
Percent Purity ≥98%
Odor Odorless
Sensitivity Hygroscopic
Density 2.00 g/mL
Chemical Name or Material Magnesium bromide hexahydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.