Learn More
Manganese(II) iodide, ultra dry, 99.99% (metals basis)
Product Code. 15497425
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15497425 1 g 1g
15407435 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15497425 Supplier Thermo Scientific Alfa Aesar Supplier No. 047166.03
 Restricted Item
Expédition estimée: 15-12-2026
Add to basket
This item is not returnable. View return policy
CAS: 7790-33-2
It is often used in the lighting industry and it is a source of its derivatives.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Ampouled under argon
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7790-33-2
MDL Number MFCD00016225
Specifications
Melting Point 638°C
Color Red to White
Physical Form Beads
Quantity 1 g
Odor Odorless
Packaging Ampule
Sensitivity Air, light, and moisture sensitive
Density 5.0 g/mL
Merck Index 14,5732
Solubility Information Soluble in water.
Formula Weight 308.75
Percent Purity 99.99%
Assay (metals basis)
Chemical Name or Material Manganese(II) iodide, Ultra dry