Learn More
Carver™ Manual Pellet Press
Product Code. 11999996
Change view
Click to view available options
Normal Force:
40 US tons
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Normal Force Unit Size
11999996 40 US tons Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Special packaging extra freight cost. Please contact us for quotation
Regulatory: This product is under eco-contribution tax.
Product Code. 11999996 Supplier Carver™ Supplier No. 4555CE
  • ONLINE EXCLUSIVE PRICE
    46390.60€
    16383.59€ / 1 pièces
    Économisez 30007.01€
    64% de réduction
 Restricted Item
Expédition estimée: 29-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Special packaging extra freight cost. Please contact us for quotation
Regulatory: This product is under eco-contribution tax.
Carver™ Pellet Presses compact homogeneous powder into a suitable pellet sample.
  • Evacuation port for connection to vacuum source
  • Oversize pressure gauge for ease of viewing
  • Inegral safety shields
Specifications
Product Type Pellet Press
For Use With (Application) Sample Prep
Includes 31 mm die
Normal Force 40 US tons