Learn More
Description
Achieve reliable and accurate results from testing with the Thermo Scientific™ MAS™ Serum Tox Control Set. This control set is used as a consistent test sample of known concentration for monitoring assay conditions in the semi-quantitative measurement of Benzodiazepine and Barbiturates in human serum and plasma, and Tricyclic Antidepressant drugs (TCA) in human serum, plasma and urine.
Ordering Information:
Thermo Scientific MAS Tox Control, Multi-Pack, 3 Levels: 2 bottles of each level, 5mL each level.
Specifications
Specifications
| Content And Storage | 2°C to 8°C |
| Description | Serum Tox Control Set |
| Detectable Analytes | Secobarbital, Diazepam, Tricyclic Antidepressants (Nortriptyline) |
| Sample Type | Plasma, Serum |
| Type | Control |
| Level | 3 |
| Quantity | 6 x 5 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.