missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ MAS™ UrichemTRAK Controls

Product Code. 17748139 Shop All Thermo Scientific Products
Change view
Click to view available options
Description:
UrichemTRAK Controls, Assayed, Bi-level
Level:
Bi-level
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Level Unit Size
17748139 UrichemTRAK Controls, Assayed, Bi-level Bi-level Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17748139 Supplier Thermo Scientific™ Supplier No. URMP

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Monitor the most commonly tested urine chemistry analytes with liquid, assayed Thermo Scientific™ MAS™ UrichemTRAK Controls, available in a convenient vial format.

Monitor the most commonly tested urine chemistry analytes with liquid, assayed Thermo Scientific™ MAS™ UrichemTRAK Controls, available in a convenient Vista™ vial format. Ready-to-use and requiring no additional preparation, these controls will not interfere with ISEs.

  • Liquid, human urine matrix
  • Open and use; no equilibration required
  • Two-year open-vial stability
  • Individual levels or multi-pack configurations
  • Stable for 24 months at 2° to 8°C
  • Assayed values available for most major analyzers
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Description UrichemTRAK Controls, Assayed, Bi-level
Detectable Analytes Amylase, Calcium, Chloride, Cortisol, Creatinine, Glucose, hCG, Magnesium, Microalbumin, Osmolality, pH, Phosphorus, Potassium, Sodium, Specific Gravity, Total Protein, Urea Nitrogen, Uric Acid
Sample Type Urine
Type Control
Level Bi-level
Packaging 6 pack - 3 bottles of each level
Quantity 15 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.