Learn More
Eppendorf™ Mastercycler Nexus GX2e, EU-Plug
Product Code. 15775675 Shop All Eppendorf Products
Change view
Click to view available options
Capacity: 64/32 x 0.2 mL PCR tubes or 24/10 x 0.5 mL PCR tube positions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15775675 Capacity: 64/32 x 0.2 mL PCR tubes or 24/10 x 0.5 mL PCR tube positions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Product Code. 15775675 Supplier Eppendorf™ Supplier No. 6338000012
En stock
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
PCR cycler with the capacity to run two simultaneous and independent protocols.
  • Large block for large assays - small block for small assays
  • Intuitive graphic programming
  • Universal block for strips, 0.1 mL, 0.2 mL, and 0.5 mL PCR tubes
  • Small footprint
  • Optional gradient for PCR optimization
  • E-mail notification
  • flexlid™ concept allows use of all types of consumables with automatic height adjustment of the lid
TRUSTED_SUSTAINABILITY
Specifications
Temperature Range (Metric) 37 to 110°C (Lid), 4 to 99°C (Block)
Content And Storage EU Plug
Dimensions 25 × 41.2 × 33 cm
Capacity 64/32 × 0.2 mL PCR Tubes or 24/10 × 0.5 mL PCR Tube Positions
Electrical Requirements 230 V, 50/60 Hz
For Use With (Application) Standard PCR, Optimizing a PCR
User Interface CAN_in/CAN_out, Ethernet
No. of Programs >700
Heated Lid Yes
Weight 10.5 kg