Learn More
Masterflex™ I/P™ Digital Modular Drive with Benchtop Controller and Remote Input/Output
Product Code. 13447099 Shop All Masterflex Products
Change view
Click to view available options
Flow Rate:
19,000 mL/min.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Flow Rate Unit Size
13447099 19,000 mL/min. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Product Code. 13447099 Supplier Masterflex™ Supplier No. 0759120
Expédition estimée: 04-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Place motor and controller where convenient with modular design
  • Flow rate 0.04 to 19 LPM with I/P™ tubing—flow rate depends on drive rpm, pump head, and tubing size
  • Digital interface with LED display shows pump rpm— confirm speed setting at a glance
  • Remote control available using analog communication—voltage, current and contact closure
  • Powerful motor lets you stack two pump heads for increased flow rate while offering ±0.25% speed control accuracy
Specifications
Electrical Requirements 115/230 VAC 149 w
Flow Rate 19,000 mL/min.
Includes 6 ft. (1.8-m) line cord with IEC 320/CEE22 socket
Overall Height (Metric) 15.2 cm
Overall Length (Metric) 39.5 cm
Overall Width (Metric) 14.1 cm
Reversible Yes
Speed Control 0.25%
Wattage 149 w
Speed 6 to 650 rpm