Learn More
Masterflex™ L/S™ Analog Modular Drive Replacement Motor
Product Code. 11751019 Shop All Masterflex Products
Change view
Click to view available options
Speed:
600 rpm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Speed Unit Size
11751019 600 rpm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Product Code. 11751019 Supplier Masterflex™ Supplier No. 0755379
Expédition estimée: 15-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Make the most of your Masterflex™ drives replacement parts and other accessories
Choose replacement motors based on your Masterflex™ Modular System
Specifications
Type Replacement Parts
Vacuum 230 VAC
Speed 600 rpm