Learn More
IKA™ Maxi MR 1 Digital
Product Code. 16208631 Shop All IKA Products
Change view
Click to view available options
Capacity:
150 L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity Unit Size
16208631 150 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Product Code. 16208631 Supplier IKA™ Supplier No. 0025002978
Equipment and Instrument End of Year Sale
Use promo code 28000
View all promotions
Expédition estimée: 11-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Magnetic stirrer without heating.
  • Powerful magnetic stirrer without heating function
  • Stirring quantity up to 150 L (H2O)
  • Flat, sturdy stainless steel casing
  • Non-locking motor
  • Infinitely variable speed
  • Digital LED speed display
  • Timer (0 to 56 min) or continuous operation
  • Integrated USB and RS 232 interface
TRUSTED_SUSTAINABILITY
Specifications
Speed 0 to 600 rpm
Capacity 150 L
Dimensions (L x W x H) 505 x 110 x 585 mm
Top Plate Surface Area (Metric) 500 x 500 mm
Electrical Requirements 50/60 Hz, 35 W
Color Blue, Silver
Timer 0 to 56 min.
Weight (Metric) 21.36 kg