Learn More
Amarell™ Permanent Magnet for Swinging Thermometer
Product Code. 10212171
Change view
Click to view available options
Immersion Type:
Total or Partial
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Immersion Type Unit Size
10212171 Total or Partial Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10212171 Supplier Amarell™ Supplier No. G13800
  • ONLINE EXCLUSIVE PRICE
    10.63€
    4.17€ / 1 pièces
    Économisez 6.46€
    60% de réduction
Expédition estimée: 29-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Use with maximum thermometers for swinging applications. The permanent magnet can be used with total or partial immersions.
  • With index and magnet
  • Use with total and partial immersion
Specifications
Immersion Type Total or Partial
Includes Index and magnet