Learn More
Lovibond™ MD200 3-in-1 photometer kit for chlorine pH and bromine
Product Code. 12681715
Change view
Click to view available options
Product Type:
Photometer
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
12681715 Photometer Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Product Code. 12681715 Supplier Lovibond™ Supplier No. 2861802
  • ONLINE EXCLUSIVE PRICE
    715.00€
    631.77€ / 1 pièces
    Économisez 83.23€
    11% de réduction
View all promotions
Expédition estimée: 15-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
MD200 3-in-1 photometer kit for chlorine pH and bromine with tablet reagents includes tablet reagents 3 round vials with lids carry case batteries and instruction manual
Versatile, compact and combination of high-quality interference filters with long-term stable LEDs as light sources in a high-precision optic results in precise measurement results at all times.
Versatile use
The MD 200 photometer can be used in almost all areas of water analysis.
Precise and low maintenance
The high-precision optics, which in addition to long-term stable LEDs as light sources, also offer high-quality interference filters, ensure precise and reproducible measurement results. In addition, these components make the photometer low-maintenance.
User friendly
The devices impress with their ease of use, ergonomic design and compact dimensions, which makes handling the devices easy and safe.
Reagents
Use long-term stable Lovibond™ reagent tablets (5-10 years guaranteed minimum durability) or Lovibond™ liquid reagents.
Specifications
Product Type Photometer
Display Type LCD
For Use With (Application) Chlorine, pH and Bromine Measurement