Learn More
Megestrol Acetate, TRC
Product Code. 30012329
Change view
Click to view available options
Quantity:
50 mg
100 mg
1 g
Unit Size:
100mg
1g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30012329 50 mg 50mg
30137836 1 g 1g
30003423 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012329 Supplier Toronto Research Chemicals Supplier No. TRCM20805050MG
 Restricted Item
Expédition estimée: 10-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 595-33-5
Molecular Formula C24 H32 O4
Molecular Weight (g/mol) 384.51
Synonym Megestrol Acetate, 6-Methyl-3,20-dioxopregna-4,6-dien-17-yl Acetate, Medroxyprogesterone Acetate Imp. G (EP), 17-(Acetyloxy)-6-methylpregna-4,6-diene-3,20-dione, 17-Hydroxy-6-methylpregna-4,6-diene-3,20-dione acetate, 17-Acetoxy-6-methylpregna-4,6-diene-3,20-dione, 17-Hydroxy-6-methylpregna-4,6-diene-3,20-dione acetate, 17α-Acetoxy-6-dehydro-6-methylprogesterone, 17α-Acetoxy-6-methylpregna-4,6-diene-3,20-dione, 17α-Hydroxy-6-methylpregna-4,6-diene-3,20-dione acetate, 5071, 6-Dehydro-6-methyl-17α-acetoxyprogesterone, 6-Methyl-17α-acetoxy-4,6-pregnadiene-3,20-dione, 6-Methyl-17α-hydroxy-δ6-progesterone acetate, 6-Methyl-6-dehydro-17α-acetoxyprogesterone, 6-Methyl-δ4,6-pregnadien-17α-ol-3,20-dione acetate, BDH 1298, DMAP, MGA, Magestin, Maygace, Megace, Megalia, Megeron, Megestat, Megestil, Megestin, Megestrol acetate, Megestryl acetate, NSC 71423, Nia, Niagestin, Ovaban, Ovarid, SC 10363
IUPAC Name [(8R,9S,10R,13S,14S,17R)-17-acetyl-6,10,13-trimethyl-3-oxo-2,8,9,11,12,14,15,16-octahydro-1H-cyclopenta[a]phenanthren-17-yl] acetate
SMILES CC(=O)O[C@@]1(CC[C@H]2[C@@H]3C=C(C)C4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C)C(=O)C
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C24H32O4/c1-14-12-18-19(22(4)9-6-17(27)13-21(14)22)7-10-23(5)20(18)8-11-24(23,15(2)25)28-16(3)26/h12-13,18-20H,6-11H2,1-5H3/t18-,19+,20+,22-,23+,24+/m1/s1
Formula Weight 384.2301
Percent Purity >95%
Chemical Name or Material Megestrol-17-acetate
Grade HPLC