Learn More
BUCHI Melting Point Sample Loader M-569
Product Code. 17815092 Shop All Buchi Products
Change view
Click to view available options
Height (Metric):
62 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Height (Metric) Unit Size
17815092 62 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17815092 Supplier BUCHI Supplier No. 051997
 Restricted Item
Expédition estimée: 07-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Fast and efficient melting point sample loading
  • Prepare 3 samples in melting point capilaries at one time
  • This unique vibration system for loading of melting point capillaries
  • Obtain reproducible results with homogeneous packing
TRUSTED_SUSTAINABILITY
Specifications
Product Type Melting Point System
Electrical Requirements 100/240 V
Height (Metric) 62 mm