Learn More
Mestranol, Mikromol™
Product Code. 18061490
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18061490 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18061490 Supplier Mikromol Supplier No. MM1698.00
 Restricted Item
Expédition estimée: 30-11-2026
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Mestranol
TRUSTED_SUSTAINABILITY
Spécification
Chemical Name or Material Mestranol
CAS 72-33-3
Analyte or Component Names Mestranol
Shipping Condition Room Temperature
Molecular Formula C21 H26 O2
Certifications/Compliance ISO 17025
Synonym Mestranol, 17-Ethynylestradiol 3-methyl ether, 3-Methoxyethynylestradiol, 3-Methylethynylestradiol, 3-O-Methylethynylestradiol, 33355, 8027 C. B., Devocin, EE 3ME, Ethinylestradiol 3-methyl ether, Ethynylestradiol 3-methyl ether, Ethynylestradiol 3-monomethyl ether, Ethynylestradiol methyl ether, Inostral, Menophase, NSC 84032, Norquen, Ovastol, SC 4725, 3-methoxy-19-nor-17alpha-pregna-1,3,5(10)-trien-20-yn-17-ol, 19-Nor-17alpha-pregna-1,3,5(10)-trien-20-yn-17-ol, 3-methoxy- (6CI,8CI), 17-Ethynyl-3-methoxy-1,3,5(10)-estratrien-17beta-o1, 17-Ethynyl-3-methoxyestra-1,3,5(10)-trien-17beta-ol, 17alpha-Ethinylestradiol 3-methyl ether, 17alpha-Ethynyl-3-methoxy-1,3,5(10)-estratrien-17beta-ol, 17alpha-Ethynyl-3-methoxy-17beta-hydroxy-Delta1,3,5(10)-estratriene, 17alpha-Ethynylestradiol 3-methyl ether, 17alpha-Ethynylestradiol methyl ether, 3-Methoxy-17alpha-ethinylestradiol, 3-Methoxy-17alpha-ethynylestradiol, 3-Methoxy-19-nor-17alpha-pregna-1,3,5(10)-trien-20-yn-17-ol, 3-Methoxy-19-nor-17alpha-pregna-1,3,5(10)-trien-20-yn-17beta-ol, 3-O-Methyl-17alpha-ethynylestradiol, delta-MVE, 19-Norpregna-1,3,5(10)-trien-20-yn-17-ol, 3-methoxy-, (17alpha)-
InChI Formula InChI=1S/C21H26O2/c1-4-21(22)12-10-19-18-7-5-14-13-15(23-3)6-8-16(14)17(18)9-11-20(19,21)2/h1,6,8,13,17-19,22H,5,7,9-12H2,2-3H3/t17-,18-,19+,20+,21+/m1/s1
SMILES COc1ccc2[C@H]3CC[C@@]4(C)[C@@H](CC[C@@]4(O)C#C)[C@@H]3CCc2c1
IUPAC Name (8R,9S,13S,14S,17R)-17-ethynyl-3-methoxy-13-methyl-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthren-17-ol
Molecular Weight (g/mol) 310.43
Quantity 250 mg
Formula Weight 310.1933 g/mol