Learn More
Beckman Coulter Life Sciences MET ONE HHPC 2+ Handheld
Product Code. 31777222 Shop All Beckman Coulter Life Sciences Products
Change view
Click to view available options
Particle Size:
0.5 to 5 μm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Particle Size Unit Size
31777222 0.5 to 5 μm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Product Code. 31777222 Supplier Beckman Coulter Life Sciences Supplier No. 208931003
Spotlight Offers
Use promo code 28038
View all promotions
Expédition estimée: 14-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Designed as an affordable tool for the routine monitoring of controlled environments to Cleanroom Classifications.
  • Designed for regulated and semi regulated production environments adhering to GMP principles
  • Suitable for aseptically packaged food and beverage filling lines
  • Used in hospital labs and formulating pharmacies
  • MET ONE HHPC 2+ is frequently chosen for these applications
Specifications
Product Type Particle Counter
Display Type TFT Color LCD
Particle Size 0.5 to 5 μm
Designed for spot checks and troubleshooting; not intended to replace continuous cleanroom monitoring systems.