Learn More
Metal Additives Standard, MA5: Ba, Ca, Mg, P, Zn @ 900 μg/g in Hydrocarbon Oil, VHG™
Product Code. 17914316
Change view
Click to view available options
Quantity:
50 g
100 g
200 g
400 g
Unit Size:
100g
200g
400g
50g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17914316 400 g 400g
17964396 100 g 100g
17972836 200 g 200g
17970486 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 17914316 Supplier VHG Labs Supplier No. VHGMA5900400G
 Restricted Item
Expédition estimée: 17-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Metal Additives Standard, MA5: Ba, Ca, Mg, P, Zn @ 900 μg/g in Hydrocarbon Oil
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Barium/Zinc/Phosphorus/Calcium/Magnesium
CAS 7440-39-3, 7440-66-6, 7723-14-0, 7440-70-2, 7439-95-4
Methods ICP/XRF/RDE
Analyte or Component Names Barium/Zinc/Phosphorus/Calcium/Magnesium
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix 75 cSt Hydrocarbon Oil
Quantity 400 g
Concentration or Composition (by Analyte or Components) Barium 900 μg/g/Zinc 900.1 μg/g/Phosphorus 900.3 μg/g/Calcium 900.4 μg/g/Magnesium 900.4 μg/g
Produkttitel
Vælg et problem
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.