Learn More
Sampling Systems™ Metal Detectable Scrapers
Product Code. 17554716 Shop All Sampling Systems Products
Change view
Click to view available options
Color:
Blue
Unit Size:
Pack of 4
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
17554716 Blue Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17554716 Supplier Sampling Systems™ Supplier No. M1019B110
  • ONLINE EXCLUSIVE PRICE
    24.71€
    21.29€ / 4 pièces
    Économisez 3.42€
    13% de réduction
Expédition estimée: 29-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Metal Detectable Scrapers are ideal for working with powders, pastes, gels and granules in production, industrial and laboratories.
  • They are made from a special chemically resistant grade of PP.
  • The whole range is manufactured in cleanroom conditions and packed in a class 7 (10,000) cleanroom.
  • This ensures the cleanliness of the product.
  • The robust design means they can be used for years and years or used as a single use product straight from the bag.
  • Simply unwrap and use.
  • Our ergonomic handle means they are easy to use.
  • Made from high quality PP.
  • Conforms to the latest FDA and EU food requirements.
  • 110 mm blade width is a standard size.
Specifications
Color Blue
Material Polypropylene
Width (Metric) 110 mm