missing translation for 'onlineSavingsMsg'
Learn More

Methallyl phenyl ether, 96%

Product Code. 11420467
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11420467

Brand: Thermo Scientific Alfa Aesar B22369.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5820-22-4 | C10H12O | 148.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5820-22-4
Molecular Formula C10H12O
Molecular Weight (g/mol) 148.21
MDL Number MFCD00008597
InChI Key LECDNXOCIPRJNJ-UHFFFAOYSA-N
Synonym methallyl phenyl ether, 2-methylallyloxy benzene, ether, 2-methylallyl phenyl, benzene, 2-methyl-2-propenyl oxy, 2-methylallyl phenyl ether, 2-methylprop-2-en-1-yl oxy benzene, 2-methyl-2-propen-1-yl oxy benzene, phenyl-methallyl ether, acmc-1b20w
PubChem CID 79893
IUPAC Name 2-methylprop-2-enoxybenzene
SMILES CC(=C)COC1=CC=CC=C1

Specifications

Density 0.964
Boiling Point 204°C
Flash Point 77°C (171°F)
Refractive Index 1.515
Quantity 25 g
Beilstein 1933868
Formula Weight 148.21
Percent Purity 96%
Chemical Name or Material Methallyl phenyl ether

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.