Learn More
Methyl 2-bromohexanoate, 95%, BLD Pharm™
Product Code. 31006989
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
Unit Size:
100g
1kg
25g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31006989 25 g 25g
30989699 100 g 100g
30996201 500 g 500g
30989523 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 31006989 Supplier BLD Pharm Supplier No. BD13821225g
 Restricted Item
Expédition estimée: 27-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
CAS 5445-19-2
Molecular Formula C7H13BrO2
MDL Number MFCD00045038
Quantity 25 g
SMILES CCCCC(Br)C(OC)=O
Molecular Weight (g/mol) 209.08
Percent Purity 95%
Chemical Name or Material Methyl 2-bromohexanoate