Learn More
Methyl 4-Amino-2-hydroxybenzoate, Mikromol™
Product Code. 30050052
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30050052 100 mg 100mg
30050957 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30050052 Supplier Mikromol Supplier No. MM0073.09
 Restricted Item
Expédition estimée: 23-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Methyl 4-Amino-2-hydroxybenzoate
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Methyl 4-Amino-2-hydroxybenzoate
CAS 4136-97-4
Analyte or Component Names Methyl 4-Amino-2-hydroxybenzoate
Shipping Condition Room Temperature
Molecular Formula C8 H9 N O3
Certifications/Compliance ISO 9001
Synonym Methyl 4-Amino-2-hydroxybenzoate, 4-Amino-2-hydroxybenzoic acid methyl ester, Salicylic acid, 4-amino-, methyl ester (6CI,7CI,8CI), 2-Hydroxy-4-aminobenzoic acid methyl ester, 3-Hydroxy-4-methoxycarbonylaniline, 4-Amino-2-hydroxybenzoic acid methyl ester, 4-Aminosalicylic acid methyl ester, Methyl 2-hydroxy-4-aminobenzoate, Methyl 4-amino-2-hydroxybenzoate, Methyl 4-aminosalicylate, Methyl p-aminosalicylate, NSC 30263, NSC 48927, p-Aminosalicylic acid methyl ester
InChI Formula InChI=1S/C8H9NO3/c1-12-8(11)6-3-2-5(9)4-7(6)10/h2-4,10H,9H2,1H3
SMILES COC(=O)c1ccc(N)cc1O
IUPAC Name methyl 4-amino-2-hydroxybenzoate
Molecular Weight (g/mol) 167.16
Quantity 100 mg
Formula Weight 167.0582 g/mol