Learn More
Methyl 8-chloro-8-oxooctanoate, 98%, BLD Pharm™
Product Code. 30979612
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
25 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30979612 250 mg
30977675 1 g
30974159 5 g
30979472 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 30979612 Supplier BLD Pharm Supplier No. BD139303250mg
 Restricted Item
Expédition estimée: 27-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
CAS 41624-92-4
Molecular Formula C9H15ClO3
MDL Number MFCD00134649
Quantity 250 mg
SMILES O=C(OC)CCCCCCC(Cl)=O
Molecular Weight (g/mol) 206.67
Percent Purity 98%
Chemical Name or Material Methyl 8-chloro-8-oxooctanoate