Learn More
Methyl benzo[b]thiophene-2-carboxylate, 98%, Thermo Scientific™
Product Code. 11444824
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11444824 25 g 25g
11424824 1 g 1g
11434824 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11444824 Supplier Thermo Scientific Alfa Aesar Supplier No. A13769.14
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This item is not returnable. View return policy
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 22913-24-2
Molecular Formula C10H8O2S
Molecular Weight (g/mol) 192.23
MDL Number MFCD00067791
InChI Key KRRAZMUPVIGDCU-UHFFFAOYSA-N
Synonym methyl benzo b thiophene-2-carboxylate, benzo b thiophene-2-carboxylic acid methyl ester, benzo b thiophene-2-carboxylic acid, methyl ester, methylbenzo b thiophene-2-carboxylate, maybridge1_001812, methyl thianaphthene-2-carboxylate, benzo b thiophene-2-carboxylicacid, methyl ester
PubChem CID 2800344
IUPAC Name methyl 1-benzothiophene-2-carboxylate
SMILES COC(=O)C1=CC2=CC=CC=C2S1
Specifications
Melting Point 69°C to 70°C
Quantity 25 g
Beilstein 142166
Formula Weight 192.24
Percent Purity 98%
Chemical Name or Material Methyl benzo[b]thiophene-2-carboxylate