Learn More
methyl N-(3,5-dichlorophenyl)carbamate, Thermo Scientific™
Product Code. 11360857
Change view
Click to view available options
Quantity:
10 g
25 g
50 g
Unit Size:
10g
25g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11360857 50 g 50g
10087632 10 g 10g
10255384 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11360857 Supplier Thermo Scientific Maybridge Supplier No. TL00611EE
 Restricted Item
Expédition estimée: 25-11-2026
Add to basket
This item is not returnable. View return policy
Chemical Identifiers
CAS 25217-43-0
Molecular Formula C8H7Cl2NO2
Molecular Weight (g/mol) 220.05
MDL Number MFCD00126402
InChI Key FRSRGACXHCLBTC-UHFFFAOYSA-N
Synonym methyl 3,5-dichlorophenyl carbamate, methyl n-3,5-dichlorophenyl carbamate, mdpc, methyl 3,5-dichlorocarbanilate, 3,5-dichlorocarbanilic acid methyl ester, carbamic acid, 3,5-dichlorophenyl-, methyl ester 9ci, carbanilic acid, 3,5-dichloro-, methyl ester, n-3,5-dichlorophenyl methoxycarboxamide, maybridge1_000169
PubChem CID 32842
IUPAC Name methyl N-(3,5-dichlorophenyl)carbamate
SMILES COC(=O)NC1=CC(Cl)=CC(Cl)=C1
Specifications
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 50 g
Formula Weight 220.05
Percent Purity 97%
Chemical Name or Material Methyl N-(3, 5-dichlorophenyl)carbamate