Learn More
Methyl perfluoro-2,5-dimethyl-3,6-dioxanonanoate, 97%, Thermo Scientific™
Product Code. 11369255
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11369255 5 g 5g
11359255 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11369255 Supplier Thermo Scientific Alfa Aesar Supplier No. L16586.06
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This item is not returnable. View return policy
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Identificadores de productos químicos
CAS 26131-32-8
Molecular Formula C10H3F17O4
Molecular Weight (g/mol) 510.103
MDL Number MFCD00054723
InChI Key UMADKHFVSXRTGY-UHFFFAOYSA-N
Synonym methyl 2,3,3,3-tetrafluoro-2-1,1,2,3,3,3-hexafluoro-2-perfluoropropoxy propoxy propanoate, perfluoro-2,5-dimethyl-3,6-dioxanonanoic acid methyl ester, methyl 2,3,3,3-tetrafluoro-2-1,1,2,3,3,3-hexafluoro-2-1,1,2,2,3,3,3-heptafluoropropoxy propoxy propanoate, methyl 2,3,3,3-tetrafluoro-2-1,1,2,3,3,3-hexafluoro-2-heptafluoropropoxy propoxy propanoate, cf3cf2cf2ocf cf3 cf2ocf cf3 cooch3, methyl perfluoro-2,5-dimethyl-3,6-dioxa-nonanoate, methyl 2,5-bis trifluoromethyl-3,6-dioxaundecafluorononanoate, 2,5-bis trifluoromethyl-3,6-dioxaundecafluorononanoic acid methyl ester, 2-2-heptafluoropropoxy hexafluoropropoxy-2,3,3,3-tetrafluoropropionic acid methyl ester, methyl2,5-bis trifluoromethyl-3,6-dioxaundecafluorononanoate mixtureofisomers
PubChem CID 141268
IUPAC Name methyl 2,3,3,3-tetrafluoro-2-[1,1,2,3,3,3-hexafluoro-2-(1,1,2,2,3,3,3-heptafluoropropoxy)propoxy]propanoate
SMILES COC(=O)C(C(F)(F)F)(OC(C(C(F)(F)F)(OC(C(C(F)(F)F)(F)F)(F)F)F)(F)F)F
Especificaciones
Boiling Point 155°C to 156°C
Refractive Index <1.300
Quantity 5 g
Formula Weight 510.12
Percent Purity 97%
Chemical Name or Material Methyl perfluoro-2,5-dimethyl-3,6-dioxanonanoate
Título del producto
Seleccione un problema
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.