Learn More
WTW™ MF45/250/E Spare Measuring Bottle (Duran 50)
Product Code. 10171481
Change view
Click to view available options
Volume (Metric):
250 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
10171481 250 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10171481 Supplier WTW™ Supplier No. 209110
 Restricted Item
Expédition estimée: 22-02-2027
Add to basket
Add to basket
This item is not returnable. View return policy
WTW offers a wide variety of measuring bottles and measuring vessels with adapters to measure BSB, soil respiration, degradeability as per OECD 301F, fermentation tests, biogas and much more. They have all be tested in their respective applications and therefore support accurate measurements.
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 250 mL