Learn More
Microbiologics™ Epower™ Enterococcus faecalis, ATCC™ 29212™†
Description
Includes
Product warranty and peel-off certificate of assay
Microorganism preparations manufactured at a predetermined concentration for versatile laboratory testing requirements.
- Process controls from one hundred to over a billion CFU/preparation
- Shelf life of 10-20 months Traceable to reference cultures
Informations sur la commande
Available in vials containing 10 lyophilized pellets, with a range of concentration from 102 to 109 per pellet
Spécification
Spécification
| ATCC® | 29212 |
| Biosafety Level | 2 |
| Form | Lyophilized Pellet |
| Product Type | Epower |
| For Use With (Application) | Lyophilized pellets add convenience to special projects and internal proficiency plans. |
| Includes | Product warranty and peel-off certificate of assay |
| Mean Assay Value | 1.0-9.9E+07 CFU per pellet |
| Packaging Type | Vial |
| Quantity | 10/Pk. |
| Species | Enterococcus faecalis |
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. Microbiologics Inc. is licensed to use these trademarks and to sell products derived from ATCC™ cultures.
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.