Learn More
Microbiologics™ Epower™ Klebsiella pneumoniae subsp. Pneumoniae, ATCC™ 10031™†
Description
Includes
Product warranty and peel-off certificate of assay
- Biosafety level: 2
- Mean Assay Value: 1.0-9.9E+07 CFU per pellet
- Methyl Red positive, Voges-Proskauer negative
Order Info
Available in vials containing 10 lyophilized pellets, with a range of concentration from 102 to 109 per pellet
Specifications
Specifications
| ATCC® | 10031 |
| Biosafety Level | 2 |
| Form | Lyophilized Pellet |
| Product Type | QC Microorganism |
| For Use With (Application) | Lyophilized pellets add convenience to special projects and internal proficiency plans. |
| Includes | Product warranty and peel-off certificate of assay |
| Mean Assay Value | 1.0-9.9E+07 CFU per pellet |
| Packaging Type | Vial |
| Quantity | 10/Pk. |
| Species | Klebsiella pneumoniae |
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. Microbiologics Inc. is licensed to use these trademarks and to sell products derived from ATCC™ cultures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.