Learn More
Microbiologics™ EZ-CFU™ One-Step Microorganism Preparations
Product Code. 11952745
Change view
Click to view available options
Species:
Aspergillus brasiliensis
Bacillus subtilis subsp. spizizenii
Clostridium sporogenes
Enterococcus faecalis
Escherichia coli
Micrococcus luteus
Pseudomonas aeruginosa
Salmonella enterica subsp. enterica serovar abony
Salmonella enterica subsp. enterica serovar typhimurium
Staphylococcus aureus subsp. aureus
ATCC®:
11437
14028
16404™*
19404
6538
6633
8739
9027
ATCC 29212
ATCC 4698
NCTC 6017
Format:
EZ-CFU
Unit Size:
Pack of 20
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Species ATCC® Format Unit Size
11952745 Bacillus subtilis subsp. spizizenii 6633 EZ-CFU Pack of 20
11942745 Aspergillus brasiliensis 16404™* EZ-CFU Pack of 20
11912755 Escherichia coli 8739 EZ-CFU Pack of 20
11902755 Clostridium sporogenes 19404 EZ-CFU Pack of 20
11982745 Clostridium sporogenes 11437 EZ-CFU Pack of 20
11932755 Staphylococcus aureus subsp. aureus 6538 EZ-CFU Pack of 20
11922755 Pseudomonas aeruginosa 9027 EZ-CFU Pack of 20
11996558 Micrococcus luteus ATCC 4698 EZ-CFU Pack of 20
11927135 Enterococcus faecalis ATCC 29212 EZ-CFU Pack of 20
13486978 Salmonella enterica subsp. enterica serovar abony NCTC 6017 EZ-CFU Pack of 20
13467376 Salmonella enterica subsp. enterica serovar typhimurium 14028 EZ-CFU Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
This item is not returnable. View return policy
Product Code. 11952745 Supplier Microbiologics™ Supplier No. 0486Z
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This item is not returnable. View return policy
Includes
Product warranty and a peel-off Certificate of Assay
Lyophilized microorganism preparations deliver less than 100CFU per 0.1mL of hydrated suspension—no dilutions are required
EZ-CFU One-Step preparations are designed to provide the required microorganism challenge concentrations for tests such as Growth Promotion Tests, Microbial Limits Tests and Microbial Enumeration Tests. These ready-to-use, dependable, and cost-effective enumerated microorganism preparations allow laboratories to conveniently and confidently conquer a variety of mandated Pharmacopoeial challenges.
  • Up to 19 tests per 2mL aliquot or 190 total tests per kit can be completed
  • Produces a durable, non-foaming suspension that is ready to be added to the challenge material immediately following the hydration step (30 minutes)
  • For added convenience, a peel-off Certificate of Assay is included for quality control documentation
  • Traceable to reference cultures
Order Info
Supplied as two vials of a single assayed microorganism (10 lyophilized pellets in each vial) and 10 vials of hydrating fluid (2.0mL in each vial)
†The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. MicroBioLogics Inc. is licensed to use these trademarks and to sell products derived from ATCC™ cultures.
Specifications
ATCC® 6633
Format EZ-CFU
Product Type EZ-CFU
Includes Product warranty and peel-off certificate of assay
Packaging Type Vial
Species Bacillus subtilis subsp. spizizenii