missing translation for 'onlineSavingsMsg'
Learn More

Microbiologics™ EZ-PEC™ Microorganism Preparations

Product Code. 11971416
Encompass_Preferred
Change view
Click to view available options
Species:
Staphylococcus aureus
ATCC®:
6538
Unit Size:
Pack of 20
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Species ATCC® Unit Size
11971416 Staphylococcus aureus 6538 Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11971416 Supplier Microbiologics™ Supplier No. 0485PEC
 Restricted Item
Expédition estimée: 15-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

2 Vials of 10 pellets ea.

Add convenience to pharmaceutical and cosmetic testing labs needing to meet antimicrobial effectiveness or preservative efficacy testing

Predetermined mean assay value results in a final challenge concentration of 100,000 to 1,000,000CFU/1.0mL of product tested

  • Enumerated lyophilized microorganism preparations are designed to provide the required microorganism challenge concentrations for Antimicrobial Effectiveness and Preservative Efficacy Tests
  • Traceable to reference cultures

Order Info

Each kit contains 20 lyophilized pellets of a single enumerated strain plus ten vials of hydrating fluid (2mL each)

†The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. MicroBioLogics Inc. is licensed to use these trademarks and to sell products derived from ATCC™ cultures.

Specifications

ATCC® 6538
Format EZ-PEC
Product Type EZ-PEC
For Use With (Application) Add convenience to pharmaceutical and cosmetic testing labs needing to meet antimicrobial effectiveness or preservative efficacy testing.
Includes Product warranty and peel-off certificate of assay
Packaging Type Vial
Quantity 2 Vials of 10 pellets ea.
Species Staphylococcus aureus
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.