Learn More
Brady™ Mild Sheet Steel Portable Metal Group Lock Box
Product Code. 11751915 Shop All Brady Products
Change view
Click to view available options
Color:
Red
Yellow
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
11751915 Yellow Each
11711925 Red Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11751915 Supplier Brady™ Supplier No. 065672
  • ONLINE EXCLUSIVE PRICE
    108.00€
    85.92€ / 1 pièces
    Économisez 22.08€
    20% de réduction
En stock
Add to basket
Add to basket
This item is not returnable. View return policy
Simplifies lockout of large equipment
  • Heavy steel lock box stores keys after machine or process is locked out
  • Each authorized work team member adds a personal lock or tagout device on the group lock box
  • Ensures that no one employee has access unless all employees have removed their locks or tags
  • Accepts up to 13 individual locks
Specifications
Product Type Portable Metal Group Lock Box
Alternate Part Number STEEL PORTABLE GROUP LOCK BOX YLW
Color Yellow
Quantity 1 Ea.